Product Details

SNP ID
rs12756167
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:110363926 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGAATGATTTCTGTTAGGGAAG[A/G]AAATTTTGCCATTAGTAACTCTCAG
Phenotype
MIM: 606077 MIM: 603878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM15 PubMed Links

Gene Details

Gene
RBM15
Gene Name
RNA binding motif protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201545.1 Intron NP_001188474.1
NM_022768.4 Intron NP_073605.4
Gene
SLC16A4
Gene Name
solute carrier family 16 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201546.1 Intron NP_001188475.1
NM_001201547.1 Intron NP_001188476.1
NM_001201548.1 Intron NP_001188477.1
NM_001201549.1 Intron NP_001188478.1
NM_001319220.1 Intron NP_001306149.1
NM_004696.2 Intron NP_004687.1
XM_005271317.4 Intron XP_005271374.1
XM_006711033.3 Intron XP_006711096.1

View Full Product Details