Product Details

SNP ID
rs13380828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42296437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAAATTAGCTTGAAAGGAACTA[A/T]CCCCAGGTACAATTTGAGGAGTCAT
Phenotype
MIM: 601511
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STAT5A PubMed Links

Gene Details

Gene
STAT5A
Gene Name
signal transducer and activator of transcription 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288718.1 Intron NP_001275647.1
NM_001288719.1 Intron NP_001275648.1
NM_001288720.1 Intron NP_001275649.1
NM_003152.3 Intron NP_003143.2
XM_005257624.3 Intron XP_005257681.1

View Full Product Details