Product Details
- SNP ID
-
rs10883875
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103500160 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGAACCCAGAGCACCCATCCTGCA[C/T]GCCCTGTTGGTTTTCAGTTCGGTTT
- Phenotype
-
MIM: 603804
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NEURL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74395445] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEURL1
- Gene Name
- neuralized E3 ubiquitin protein ligase 1
- Gene
- NEURL1-AS1
- Gene Name
- NEURL1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details