Product Details
- SNP ID
-
rs12259605
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:90867822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGAATGGAATAAACTTAGGATAA[A/G]GCCAAAGACTAATCAAGCACACACC
- Phenotype
-
MIM: 182137
MIM: 606115
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HTR7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112495378] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTR7
- Gene Name
- 5-hydroxytryptamine receptor 7
There are no transcripts associated with this gene.
- Gene
- RPP30
- Gene Name
- ribonuclease P/MRP subunit p30
View Full Product Details