Product Details

SNP ID
rs13439542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22620505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCCTCCACAGCTCTTCCTACTC[C/T]CCTAGGCTTGCCCGCGTGCCCTCCC
Phenotype
MIM: 606396 MIM: 607359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIN3 PubMed Links

Gene Details

Gene
BIN3
Gene Name
bridging integrator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018688.4 1776 UTR 3 NP_061158.1
XM_005273573.3 1776 Intron XP_005273630.1
XM_011544586.2 1776 Intron XP_011542888.1
XM_011544587.1 1776 Intron XP_011542889.1
Gene
CCAR2
Gene Name
cell cycle and apoptosis regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021174.5 1776 Intron NP_066997.3
XM_011544603.1 1776 Intron XP_011542905.1
XM_011544604.1 1776 Intron XP_011542906.1
XM_017013717.1 1776 Intron XP_016869206.1

View Full Product Details