Product Details
- SNP ID
-
hCV31680044
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:11559257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGGCTCAGGGGCGAGAGGTTTCA[G/T]CCAACAGCATGACAGAGAAGGGCTC
- Phenotype
-
MIM: 608760
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATG7
PubMed Links
Gene Details
- Gene
- ATG7
- Gene Name
- autophagy related 7
There are no transcripts associated with this gene.
- Gene
- VGLL4
- Gene Name
- vestigial like family member 4
There are no transcripts associated with this gene.
View Full Product Details