Product Details

SNP ID
rs12106668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12291996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAATTTAATTGAGCTGATATT[C/G]TGAAAAGAACCTTCATGAATATCTA
Phenotype
MIM: 601487
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPARG PubMed Links

Gene Details

Gene
PPARG
Gene Name
peroxisome proliferator activated receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005037.5 Intron NP_005028.4
NM_015869.4 Intron NP_056953.2
NM_138711.3 Intron NP_619725.2
NM_138712.3 Intron NP_619726.2
XM_011533841.2 Intron XP_011532143.1
XM_011533842.2 Intron XP_011532144.1
XM_011533843.2 Intron XP_011532145.1
XM_011533844.1 Intron XP_011532146.1

View Full Product Details