Product Details

SNP ID
rs11129823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39386750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGTTTTGGAGATAGAGTGAA[C/T]AAGTGTGGAGAGGCCAAGTCCAAGG
Phenotype
MIM: 610819
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A38 PubMed Links

Gene Details

Gene
SLC25A38
Gene Name
solute carrier family 25 member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017875.2 190 Intron NP_060345.2
XM_006713214.1 190 Intron XP_006713277.1
XM_011533869.2 190 UTR 5 XP_011532171.1
XM_017006710.1 190 Intron XP_016862199.1

View Full Product Details