Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001165928.3 | 394 | UTR 5 | NP_001159400.2 | ||
NM_001177634.2 | 394 | UTR 5 | NP_001171105.1 | ||
NM_001177635.2 | 394 | Intron | NP_001171106.1 | ||
NM_001177636.2 | 394 | UTR 5 | NP_001171107.1 | ||
NM_001177637.2 | 394 | Intron | NP_001171108.1 | ||
NM_001177638.2 | 394 | Intron | NP_001171109.1 | ||
NM_001177639.2 | 394 | Intron | NP_001171110.1 | ||
NM_001177640.2 | 394 | Intron | NP_001171111.1 | ||
NM_001177641.2 | 394 | Intron | NP_001171112.1 | ||
NM_001177642.2 | 394 | Intron | NP_001171113.1 | ||
NM_001177643.2 | 394 | Intron | NP_001171114.1 | ||
NM_001177644.2 | 394 | Intron | NP_001171115.1 | ||
NM_004393.5 | 394 | Intron | NP_004384.4 |