Product Details
- SNP ID
-
rs13065840
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51969419 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCAGCCCACCCCCTGCAGCAGTG[A/C]TGGGCTTCACTGGAGCCCCTGCAGG
- Phenotype
-
MIM: 608503
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD14A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112901597] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABHD14A
- Gene Name
- abhydrolase domain containing 14A
There are no transcripts associated with this gene.
- Gene
- ABHD14A-ACY1
- Gene Name
- ABHD14A-ACY1 readthrough
There are no transcripts associated with this gene.
- Gene
- ABHD14B
- Gene Name
- abhydrolase domain containing 14B
- Gene
- PCBP4
- Gene Name
- poly(rC) binding protein 4
There are no transcripts associated with this gene.
View Full Product Details