Product Details
- SNP ID
-
rs11041276
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7243071 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATAAATAAATAAATAAATAAATA[A/T]AAAGTATTACCAATATGAACAGGAT
- Phenotype
-
MIM: 613528
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR302E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202188440] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR302E
- Gene Name
- microRNA 302e
There are no transcripts associated with this gene.
- Gene
- SYT9
- Gene Name
- synaptotagmin 9
View Full Product Details