Product Details

SNP ID
rs10775586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2867867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTGCGGGCGCTCCACGTGGCGG[C/G]ACGGGCTGGCCTGACCCCAAAGGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
There are no transcripts associated with this gene.

Gene
ZNF556
Gene Name
zinc finger protein 556
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300843.1 Intron NP_001287772.1
NM_024967.2 Intron NP_079243.1

View Full Product Details