Product Details

SNP ID
rs6429283
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:241652861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTACTTCTCTGAACC[G/T]AAGTTGCTTCAATTGTAAAACTATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR64 PubMed Links

Gene Details

Gene
WDR64
Gene Name
WD repeat domain 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144625.4 Intron NP_653226.4
XM_006711736.3 Intron XP_006711799.1
XM_011544085.2 Intron XP_011542387.1
XM_011544086.2 Intron XP_011542388.1
XM_011544087.2 Intron XP_011542389.1
XM_011544091.1 Intron XP_011542393.1
XM_011544092.2 Intron XP_011542394.1
XM_017000315.1 Intron XP_016855804.1

View Full Product Details