Product Details

SNP ID
rs12035584
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:3814033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTACATTGTTCAAATCGGGATTA[A/G]TTTTTAGATATTTTGTTGTTTCACA
Phenotype
MIM: 616690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP104 PubMed Links

Gene Details

Gene
CEP104
Gene Name
centrosomal protein 104
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014704.3 4806 UTR 3 NP_055519.1
XM_005244815.4 4806 Intron XP_005244872.1
XM_011542473.2 4806 UTR 3 XP_011540775.1
XM_011542474.2 4806 Intron XP_011540776.1
XM_011542475.2 4806 Intron XP_011540777.1
XM_011542476.2 4806 Intron XP_011540778.1
XM_011542477.2 4806 Intron XP_011540779.1
XM_011542478.1 4806 Intron XP_011540780.1
XM_017002917.1 4806 Intron XP_016858406.1
XM_017002918.1 4806 Intron XP_016858407.1
XM_017002919.1 4806 Intron XP_016858408.1

View Full Product Details