Product Details

SNP ID
rs12800499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55795609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCCCACCTTTGCCCTTTTAGG[G/T]TTCACAGATTACCCAAAGCTTCAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5D14 PubMed Links

Gene Details

Gene
OR5D14
Gene Name
olfactory receptor family 5 subfamily D member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004735.1 54 Silent Mutation GGG,GGT G18G NP_001004735.1

View Full Product Details