Product Details

SNP ID
rs12807051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60852792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGAGCGCCGCAGCTTGCGCAGCA[G/T]GTCGGGGCAGGTGAGCACGTAGAGC
Phenotype
MIM: 611293 MIM: 604837
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC86 PubMed Links

Gene Details

Gene
CCDC86
Gene Name
coiled-coil domain containing 86
There are no transcripts associated with this gene.

Gene
PTGDR2
Gene Name
prostaglandin D2 receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004778.2 1043 Missense Mutation ATG,CTG M311L NP_004769.2

View Full Product Details