Product Details

SNP ID
rs12276791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75422841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGTCCCTGCCTGGCCAGGCCA[C/G]ATAATAGAACCTTGACCCACAGACT
Phenotype
MIM: 600454
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHL35 PubMed Links

Gene Details

Gene
KLHL35
Gene Name
kelch like family member 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039548.2 Intron NP_001034637.2
XM_011544954.2 Intron XP_011543256.1
Gene
RPS3
Gene Name
ribosomal protein S3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005.4 Intron NP_000996.2
NM_001256802.1 Intron NP_001243731.1
NM_001260506.1 Intron NP_001247435.1
NM_001260507.1 Intron NP_001247436.1

View Full Product Details