Product Details
- SNP ID
-
rs12286009
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:75427765 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTCTGCATGTCCCTGCCCTTGT[A/G]CCTTTGCCCATACTGTTCCCTCCAA
- Phenotype
-
MIM: 609632
MIM: 600454
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GDPD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs377710536] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GDPD5
- Gene Name
- glycerophosphodiester phosphodiesterase domain containing 5
There are no transcripts associated with this gene.
- Gene
- KLHL35
- Gene Name
- kelch like family member 35
- Gene
- RPS3
- Gene Name
- ribosomal protein S3
There are no transcripts associated with this gene.
View Full Product Details