Product Details

SNP ID
rs11905883
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59151062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTTCCGATAGTGCAGGGGTACC[A/G]GGCTGATAGCGGCTGGGTCTCTGGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF831 PubMed Links
Additional Information
For this assay, SNP(s) [rs80271186] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ZNF831
Gene Name
zinc finger protein 831
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178457.2 Intron NP_848552.1
XM_005260272.3 Intron XP_005260329.1
XM_005260273.3 Intron XP_005260330.1
XM_006723698.3 Intron XP_006723761.1
XM_011528534.2 Intron XP_011526836.1
XM_011528536.2 Intron XP_011526838.1
XM_011528537.2 Intron XP_011526839.1
XM_011528538.2 Intron XP_011526840.1
XM_011528540.2 Intron XP_011526842.1
XM_017027642.1 Intron XP_016883131.1
XM_017027643.1 Intron XP_016883132.1
XM_017027644.1 Intron XP_016883133.1

View Full Product Details