Product Details

SNP ID
rs12053605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46684488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCACGTGGCAGCAGGAAGTGC[C/T]GAACAAGAGGGAAAAGCCCCTTATA
Phenotype
MIM: 606411 MIM: 608679
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC13A3 PubMed Links

Gene Details

Gene
SLC13A3
Gene Name
solute carrier family 13 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011554.2 3250 Intron NP_001011554.1
NM_001193339.1 3250 Intron NP_001180268.1
NM_001193340.1 3250 Intron NP_001180269.1
NM_001193342.1 3250 Intron NP_001180271.1
NM_022829.5 3250 Intron NP_073740.2
Gene
TP53RK
Gene Name
TP53 regulating kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033550.3 3250 UTR 3 NP_291028.3

View Full Product Details