Product Details

SNP ID
rs12911167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59640212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAAAACTACTGCAATAAACAA[A/G]GCCAATCTGCTACTTCTCATCCACT
Phenotype
MIM: 600519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2A2 PubMed Links

Gene Details

Gene
GTF2A2
Gene Name
general transcription factor IIA subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320929.1 Intron NP_001307858.1
NM_001320930.1 Intron NP_001307859.1
NM_004492.2 Intron NP_004483.1

View Full Product Details