Product Details

SNP ID
rs10933952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108593980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGAGCTTGAATTTCTAGTAT[A/G]GTTACGGATCTGATCATTTATTCTG
Phenotype
MIM: 608672 MIM: 610643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DZIP3 PubMed Links

Gene Details

Gene
DZIP3
Gene Name
DAZ interacting zinc finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014648.3 Intron NP_055463.1
XM_005247914.3 Intron XP_005247971.1
XM_005247915.3 Intron XP_005247972.1
XM_005247916.3 Intron XP_005247973.1
XM_005247917.3 Intron XP_005247974.1
XM_005247918.3 Intron XP_005247975.1
XM_005247919.3 Intron XP_005247976.1
XM_017007540.1 Intron XP_016863029.1
Gene
KIAA1524
Gene Name
KIAA1524
There are no transcripts associated with this gene.

View Full Product Details