Product Details
- SNP ID
-
hCV31749487
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149432447 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGCGTGCTTGGGGTTGAACGTGG[C/G]CCCGCGTTCGGGTGAGACGCAGCCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF777
PubMed Links
Gene Details
- Gene
- ZNF777
- Gene Name
- zinc finger protein 777
There are no transcripts associated with this gene.
View Full Product Details