Product Details

SNP ID
rs12101178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105328219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTAAAGGACACATGGCTTATGCC[A/G]CACCCCCTGCCCCAACCCCCGGAGT
Phenotype
MIM: 610423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PACS2 PubMed Links

Gene Details

Gene
PACS2
Gene Name
phosphofurin acidic cluster sorting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100913.2 Intron NP_001094383.2
NM_001243127.2 Intron NP_001230056.1
NM_015197.3 Intron NP_056012.2
XM_006720090.1 Intron XP_006720153.1
XM_006720091.1 Intron XP_006720154.1
XM_006720092.2 Intron XP_006720155.1
XM_017021105.1 Intron XP_016876594.1
XM_017021106.1 Intron XP_016876595.1
XM_017021107.1 Intron XP_016876596.1
XM_017021108.1 Intron XP_016876597.1
XM_017021109.1 Intron XP_016876598.1
XM_017021110.1 Intron XP_016876599.1
XM_017021111.1 Intron XP_016876600.1
XM_017021112.1 Intron XP_016876601.1

View Full Product Details