Product Details

SNP ID
rs7833720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144161320 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTTCTGGGGATTAGGGTGTGGG[A/C]AGCTTTGGGGCTATTATTCTGCTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MROH1 PubMed Links

Gene Details

Gene
MROH1
Gene Name
maestro heat like repeat family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099280.2 Intron NP_001092750.1
NM_001099281.2 Intron NP_001092751.1
NM_001288814.1 Intron NP_001275743.1
NM_032450.2 Intron NP_115826.2
XM_011517270.2 Intron XP_011515572.1
XM_011517271.2 Intron XP_011515573.1
XM_011517272.2 Intron XP_011515574.1
XM_011517273.2 Intron XP_011515575.1
XM_011517274.2 Intron XP_011515576.1
XM_011517275.2 Intron XP_011515577.1
XM_011517276.2 Intron XP_011515578.1
XM_011517277.1 Intron XP_011515579.1
XM_011517278.1 Intron XP_011515580.1
XM_011517279.2 Intron XP_011515581.1
XM_011517281.1 Intron XP_011515583.1
XM_011517282.1 Intron XP_011515584.1
XM_011517283.2 Intron XP_011515585.1
XM_011517285.1 Intron XP_011515587.1
XM_011517286.2 Intron XP_011515588.1
XM_017013802.1 Intron XP_016869291.1
XM_017013803.1 Intron XP_016869292.1
XM_017013804.1 Intron XP_016869293.1
XM_017013805.1 Intron XP_016869294.1
XM_017013806.1 Intron XP_016869295.1
XM_017013807.1 Intron XP_016869296.1

View Full Product Details