Product Details

SNP ID
rs12447426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67229422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAAGAAAATTTTATTTTGCTCCG[A/T]GCGTTCAAGGAGCTCACACACATGC
Phenotype
MIM: 606881
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318202.1 4115 UTR 3 NP_001305131.1
NM_013241.2 4115 UTR 3 NP_037373.2
XM_006721180.1 4115 UTR 3 XP_006721243.1
XM_011523043.2 4115 UTR 3 XP_011521345.1
XM_011523044.1 4115 UTR 3 XP_011521346.1
XM_011523045.2 4115 UTR 3 XP_011521347.1
Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
TMEM208
Gene Name
transmembrane protein 208
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318217.1 4115 Intron NP_001305146.1
NM_014187.3 4115 Intron NP_054906.2

View Full Product Details