Product Details
- SNP ID
-
rs12252639
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:35128854 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCCTAGTCTTGATTCTTTTTTTG[G/T]ATAAACTTTCCCTTCCTAAGTCATA
- Phenotype
-
MIM: 123812
MIM: 603135
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CREM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114736635] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CREM
- Gene Name
- cAMP responsive element modulator
- Gene
- CUL2
- Gene Name
- cullin 2
There are no transcripts associated with this gene.
View Full Product Details