Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001097590.2 | Intron | NP_001091059.1 | |||
NM_001265601.1 | Intron | NP_001252530.1 | |||
NM_006414.4 | Intron | NP_006405.2 | |||
NM_183005.4 | Intron | NP_892117.1 | |||
XM_006717363.1 | Intron | XP_006717426.1 | |||
XM_006717364.3 | Intron | XP_006717427.1 | |||
XM_011519293.1 | Intron | XP_011517595.1 | |||
XM_017015479.1 | Intron | XP_016870968.1 | |||
XM_017015480.1 | Intron | XP_016870969.1 | |||
XM_017015481.1 | Intron | XP_016870970.1 |