Product Details

SNP ID
rs12342978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:115023138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATTTGGCCCTTTGCATTTTTAT[A/G]AAGTCCTTGGGATTCCAGTGAGCAG
Phenotype
MIM: 187380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNC PubMed Links

Gene Details

Gene
TNC
Gene Name
tenascin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002160.3 Intron NP_002151.2
XM_005251972.3 Intron XP_005252029.1
XM_005251973.3 Intron XP_005252030.1
XM_005251974.3 Intron XP_005252031.1
XM_005251975.3 Intron XP_005252032.1
XM_006717096.3 Intron XP_006717159.1
XM_006717097.3 Intron XP_006717160.1
XM_006717098.3 Intron XP_006717161.1
XM_006717101.3 Intron XP_006717164.1
XM_011518625.2 Intron XP_011516927.1
XM_011518626.2 Intron XP_011516928.1
XM_011518628.2 Intron XP_011516930.1
XM_011518629.2 Intron XP_011516931.1
XM_017014678.1 Intron XP_016870167.1
XM_017014679.1 Intron XP_016870168.1
XM_017014680.1 Intron XP_016870169.1
XM_017014681.1 Intron XP_016870170.1

View Full Product Details