Product Details

SNP ID
rs11063223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4545694 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCTTATACTCCATAGCACTTTT[A/G]AGCAGCTTGAATAGGACTTTATAGT
Phenotype
MIM: 616082 MIM: 603070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf4 PubMed Links

Gene Details

Gene
C12orf4
Gene Name
chromosome 12 open reading frame 4
There are no transcripts associated with this gene.

Gene
RAD51AP1
Gene Name
RAD51 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130862.1 Intron NP_001124334.1
NM_006479.4 Intron NP_006470.1

View Full Product Details