Product Details

SNP ID
rs6814637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38976551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTGAGACCCTAAGCAGAAGATC[C/T]GGTTAAGCCACGCACAGATATCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM156 PubMed Links

Gene Details

Gene
TMEM156
Gene Name
transmembrane protein 156
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303228.1 Intron NP_001290157.1
NM_024943.2 Intron NP_079219.1
XM_005262670.3 Intron XP_005262727.1
XM_011513749.2 Intron XP_011512051.1
XM_011513750.2 Intron XP_011512052.1
XM_011513751.2 Intron XP_011512053.1
XM_011513752.2 Intron XP_011512054.1
XM_011513753.2 Intron XP_011512055.1
XM_011513754.2 Intron XP_011512056.1
XM_017008629.1 Intron XP_016864118.1

View Full Product Details