Product Details

SNP ID
rs7219860
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58243910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTGAGGGGTGGGGGTAATGGCC[A/G]AGAAAGAGGGAGACAAAAGCAGAAG
Phenotype
MIM: 150205
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LPO PubMed Links

Gene Details

Gene
LPO
Gene Name
lactoperoxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160102.1 Intron NP_001153574.1
NM_006151.2 Intron NP_006142.1
XM_011524808.2 Intron XP_011523110.1
XM_011524810.2 Intron XP_011523112.1

View Full Product Details