Product Details

SNP ID
rs12811645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:19132700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTAGATCATTTATGGCCACTT[C/T]TAAAAATAATTTTGTATCTTCGTTT
Phenotype
MIM: 607770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHA5 PubMed Links

Gene Details

Gene
PLEKHA5
Gene Name
pleckstrin homology domain containing A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143821.2 Intron NP_001137293.2
NM_001190860.2 Intron NP_001177789.2
NM_001256470.1 Intron NP_001243399.1
NM_001256787.1 Intron NP_001243716.1
NM_019012.5 Intron NP_061885.2
XM_005253400.1 Intron XP_005253457.1
XM_006719091.1 Intron XP_006719154.1
XM_006719093.1 Intron XP_006719156.1
XM_006719096.1 Intron XP_006719159.1
XM_011520714.1 Intron XP_011519016.1
XM_011520715.1 Intron XP_011519017.1
XM_011520716.1 Intron XP_011519018.1
XM_011520717.1 Intron XP_011519019.1
XM_017019495.1 Intron XP_016874984.1
XM_017019496.1 Intron XP_016874985.1
XM_017019497.1 Intron XP_016874986.1
XM_017019498.1 Intron XP_016874987.1
XM_017019499.1 Intron XP_016874988.1
XM_017019500.1 Intron XP_016874989.1
XM_017019501.1 Intron XP_016874990.1
XM_017019502.1 Intron XP_016874991.1
XM_017019503.1 Intron XP_016874992.1

View Full Product Details