Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001306084.1 | 562 | Intron | NP_001293013.1 | ||
XM_011539069.2 | 562 | Intron | XP_011537371.1 | ||
XM_011539070.2 | 562 | Intron | XP_011537372.1 | ||
XM_011539071.2 | 562 | Intron | XP_011537373.1 | ||
XM_011539072.2 | 562 | Intron | XP_011537374.1 | ||
XM_011539073.2 | 562 | Intron | XP_011537375.1 | ||
XM_011539074.2 | 562 | Intron | XP_011537376.1 | ||
XM_011539076.2 | 562 | Intron | XP_011537378.1 | ||
XM_011539077.2 | 562 | UTR 5 | XP_011537379.1 | ||
XM_011539078.2 | 562 | Intron | XP_011537380.1 | ||
XM_011539079.1 | 562 | Intron | XP_011537381.1 | ||
XM_011539081.2 | 562 | Intron | XP_011537383.1 | ||
XM_011539083.2 | 562 | Intron | XP_011537385.1 | ||
XM_011539084.2 | 562 | Intron | XP_011537386.1 | ||
XM_011539085.1 | 562 | Intron | XP_011537387.1 | ||
XM_017018866.1 | 562 | Intron | XP_016874355.1 | ||
XM_017018867.1 | 562 | Intron | XP_016874356.1 |