Product Details

SNP ID
rs12328668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24502420 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAATTATTGGTCAAACAATTAAT[G/T]AATTGGTCAAGCAAAGTGGTCCCAC
Phenotype
MIM: 602691
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NCOA1 PubMed Links

Gene Details

Gene
NCOA1
Gene Name
nuclear receptor coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003743.4 Intron NP_003734.3
NM_147223.2 Intron NP_671756.1
NM_147233.2 Intron NP_671766.1
XM_005264625.1 Intron XP_005264682.1
XM_005264626.1 Intron XP_005264683.1
XM_005264628.1 Intron XP_005264685.1
XM_017005168.1 Intron XP_016860657.1
XM_017005169.1 Intron XP_016860658.1

View Full Product Details