Product Details

SNP ID
rs6904392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34757289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTTATAGAAAGATTAAATACG[C/T]TCTTCCGGAAGCACCGACAAACAAG
Phenotype
MIM: 603522
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRPC PubMed Links

Gene Details

Gene
SNRPC
Gene Name
small nuclear ribonucleoprotein polypeptide C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003093.2 196 UTR 5 NP_003084.1

View Full Product Details