Product Details
- SNP ID
-
rs12744435
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:35587693 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAATTAACTTCTCAGATTCCTT[A/C]TCTGGTCTTCAGACAATTAGTAGAT
- Phenotype
-
MIM: 614428
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TFAP2E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112962958,rs74988880] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFAP2E
- Gene Name
- transcription factor AP-2 epsilon
View Full Product Details