Product Details

SNP ID
rs13421903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:192751226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACAGTTCTTCATATGTAGAGGA[C/T]GTTCAAATTGGCTAAGTTCCTTTTC
Phenotype
MIM: 605443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCGEM1 PubMed Links

Gene Details

Gene
PCGEM1
Gene Name
PCGEM1, prostate-specific transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details