Product Details

SNP ID
rs4252233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69365721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCGGCATGCTCTTGGGAGAAGCC[A/G]TCTTGCCAGACTCCATGATATCCGA
Phenotype
MIM: 603139 MIM: 600822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK6 PubMed Links

Gene Details

Gene
AK6
Gene Name
adenylate kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015891.1 484 Intron NP_001015891.1
NM_016283.4 484 Intron NP_057367.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
There are no transcripts associated with this gene.

Gene
TAF9
Gene Name
TATA-box binding protein associated factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015892.1 484 Missense Mutation ACG,ATG T6M NP_001015892.1
NM_003187.4 484 Intron NP_003178.1

View Full Product Details