Product Details

SNP ID
rs12136207
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:15479745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGATAGCGTGGTCACAGAAGGC[C/T]TCATGGAGAAGGCAACCTTGAGCAG
Phenotype
MIM: 609443 MIM: 609444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
There are no transcripts associated with this gene.

Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015849.2 Intron NP_056933.2

View Full Product Details