Product Details

SNP ID
rs11583933
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:160121105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGAAACTCCGTGTCCCCCATG[A/C]TGCTCAACTCACCCCTGTGCCCTGC
Phenotype
MIM: 182340
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A2 PubMed Links

Gene Details

Gene
ATP1A2
Gene Name
ATPase Na+/K+ transporting subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000702.3 Intron NP_000693.1

View Full Product Details