Product Details

SNP ID
rs11204982
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:152349566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGAAAGAACAGACATATACCC[C/T]TACTACCAAAAGAGGAGATCATGTT
Phenotype
MIM: 616284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLG-AS1 PubMed Links

Gene Details

Gene
FLG-AS1
Gene Name
FLG antisense RNA 1
There are no transcripts associated with this gene.

Gene
FLG2
Gene Name
filaggrin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014342.2 7859 UTR 3 NP_001014364.1
XM_011509531.2 7859 UTR 3 XP_011507833.1

View Full Product Details