Product Details

SNP ID
rs13305194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:18919293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCTAGGAAAAGAATCATAGACA[A/T]AAAGATGGCACTGGTGTTGGGGCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TTTY14 PubMed Links

Gene Details

Gene
TTTY14
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details