Product Details
- SNP ID
-
rs13208325
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7330051 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGACCCTCAGAGATTAAGATCAAG[C/G]TTTTTGGTATATAACATGTGAATCA
- Phenotype
-
MIM: 608304
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CAGE1
PubMed Links
Gene Details
- Gene
- CAGE1
- Gene Name
- cancer antigen 1
View Full Product Details