Product Details

SNP ID
rs7219191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18277231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAATGAGGCTGATGCTTATGGTC[G/T]GCCAAGGGTCACGACAGCTGCAAGG
Phenotype
MIM: 601243
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TOP3A PubMed Links

Gene Details

Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320759.1 Intron NP_001307688.1
NM_004618.4 Intron NP_004609.1
XM_011524001.2 Intron XP_011522303.1

View Full Product Details