Product Details

SNP ID
rs11878398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37508429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGTATCTAAAATGATAATGTAG[G/T]CTGTGTGCAGTGGTTCATGCCTGTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF793 PubMed Links

Gene Details

Gene
ZNF793
Gene Name
zinc finger protein 793
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013659.2 Intron NP_001013681.2
XM_005258927.3 Intron XP_005258984.1
XM_006723213.3 Intron XP_006723276.1
Gene
ZNF793-AS1
Gene Name
ZNF793 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details