Product Details

SNP ID
rs10748138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:40699596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAATTTAGCTGTGTTGGAATTAA[A/G]TAATGGCAAAGTTAATTTCACTTTT
Phenotype
MIM: 600016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNTN1 PubMed Links

Gene Details

Gene
CNTN1
Gene Name
contactin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256063.1 Intron NP_001242992.1
NM_001256064.1 Intron NP_001242993.1
NM_001843.3 Intron NP_001834.2
NM_175038.2 Intron NP_778203.1
XM_005268651.2 Intron XP_005268708.1
XM_006719241.2 Intron XP_006719304.1
XM_011537926.2 Intron XP_011536228.1
XM_011537927.2 Intron XP_011536229.1
XM_017018821.1 Intron XP_016874310.1
XM_017018822.1 Intron XP_016874311.1
XM_017018823.1 Intron XP_016874312.1
XM_017018824.1 Intron XP_016874313.1
XM_017018825.1 Intron XP_016874314.1
XM_017018826.1 Intron XP_016874315.1
XM_017018827.1 Intron XP_016874316.1

View Full Product Details