Product Details

SNP ID
rs13294666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32385065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCCTGGGAGTCCTAACCCCGA[A/G]GGTGTGAGGCCCGCACTGGGAGTCC
Phenotype
MIM: 100880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACO1 PubMed Links

Gene Details

Gene
ACO1
Gene Name
aconitase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278352.1 Intron NP_001265281.1
NM_002197.2 Intron NP_002188.1
XM_005251476.1 Intron XP_005251533.1
XM_011517888.1 Intron XP_011516190.1

View Full Product Details