Product Details

SNP ID
rs7019690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5639227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATTTTTCTTCTTTTATGGTGTA[A/G]TACTTATAGCTATAAATTTCTCTGT
Phenotype
MIM: 610354
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIC1 PubMed Links

Gene Details

Gene
RIC1
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135920.2 Intron NP_001129392.2
NM_001206557.1 Intron NP_001193486.1
NM_020829.3 Intron NP_065880.2
XM_005251523.3 Intron XP_005251580.1
XM_011517967.2 Intron XP_011516269.1
XM_011517968.2 Intron XP_011516270.1
XM_017014934.1 Intron XP_016870423.1
XM_017014935.1 Intron XP_016870424.1
XM_017014936.1 Intron XP_016870425.1

View Full Product Details