Product Details

SNP ID
rs9620752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27928878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAGAAAAGAAAAAAAGTCA[A/G]TTATCAACCAGGTACAATTTCTCAT
Phenotype
MIM: 606876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3199-1 PubMed Links

Gene Details

Gene
MIR3199-1
Gene Name
microRNA 3199-1
There are no transcripts associated with this gene.

Gene
MIR3199-2
Gene Name
microRNA 3199-2
There are no transcripts associated with this gene.

Gene
PITPNB
Gene Name
phosphatidylinositol transfer protein beta
There are no transcripts associated with this gene.

Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details